Fmoc-6-bromo-DL-tryptophan
Supplier: Matrix Scientific
042149-100MG
042149-1G
101854-666EA
326.15
USD
101854-666
101854-668
Fmoc-6-bromo-DL-tryptophan
Fmoc-6-bromo-DL-tryptophan
Hazard: Irritant
Formula:
C₂₆H₂₁BrN₂O₄ MW: 505.38 g/mol |
MDL Number:
MFCD09750489 CAS Number: 753504-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...