Fmoc-5-methyl-DL-tryptophan

Supplier: Matrix Scientific

042148-100MG 042148-1G
101854-662EA 300 USD
101854-662 101854-664
Fmoc-5-methyl-DL-tryptophan
Fmoc-5-methyl-DL-tryptophan

Hazard: Irritant

Formula: C₂₇H₂₄N₂O₄
MDL Number: MFCD02682375
CAS Number: 138775-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR