Fmoc-5-methoxy-DL-tryptophan

Supplier: Matrix Scientific

042146-100MG 042146-1G
101854-654EA 249.23 USD
101854-654 101854-656
Fmoc-5-methoxy-DL-tryptophan
Fmoc-5-methoxy-DL-tryptophan

Hazard: Irritant

Formula: C₂₇H₂₄N₂O₅
MW: 456.5 g/mol
MDL Number: MFCD02682367
CAS Number: 138775-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR