Fmoc-5-methoxy-2-methyl-DL-tryptophan
Supplier: Matrix Scientific
042145-100MG
042145-1G
101854-650EA
249.23
USD
101854-650
101854-652
Fmoc-5-methoxy-2-methyl-DL-tryptophan
Fmoc-5-methoxy-2-methyl-DL-tryptophan
Hazard: Irritant
Formula:
C₂₈H₂₆N₂O₅ |
MDL Number:
MFCD02682365 CAS Number: 1219357-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...