Fmoc-5-Hydroxy-L-tryptophan
Supplier: Matrix Scientific
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-hydroxy-1H-indol-3-yl)propanoic acid, N-Fmoc-5-hydroxy-L-tryptophan, Fmoc-Trp(5-OH)-OH
042144-1G
042144-5G
101854-646EA
412.31
USD
101854-646
101854-648
Fmoc-5-Hydroxy-L-tryptophan
Fmoc-5-Hydroxy-L-tryptophan
Hazard: Irritant
Formula:
C₂₆H₂₂N₂O₅ MW: 442.46 g/mol |
MDL Number:
MFCD00270744 CAS Number: 178119-94-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...