Fmoc-5-hydroxy-DL-tryptophan

Supplier: Matrix Scientific

Synonyms: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-hydroxy-1H-indol-3-yl)propanoic acid

042143-250MG 042143-1G
101854-642EA 263.08 USD
101854-642 101854-644
Fmoc-5-hydroxy-DL-tryptophan
Fmoc-5-hydroxy-DL-tryptophan

Hazard: Irritant

Formula: C₂₆H₂₂N₂O₅
MDL Number: MFCD02682363
CAS Number: 351857-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR