Fmoc-5-fluoro-DL-tryptophan

Supplier: Matrix Scientific

042142-100MG 042142-1G
101854-638EA 187.69 USD
101854-638 101854-640
Fmoc-5-fluoro-DL-tryptophan
Fmoc-5-fluoro-DL-tryptophan

Hazard: Irritant

Formula: C₂₆H₂₁FN₂O₄
MDL Number: MFCD02682359
CAS Number: 138775-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR