Fmoc-5-benzyloxy-DL-tryptophan

Supplier: Matrix Scientific

042139-250MG 042139-1G
101854-626EA 363.08 USD
101854-626 101854-628
Fmoc-5-benzyloxy-DL-tryptophan
Fmoc-5-benzyloxy-DL-tryptophan

Hazard: Irritant

Formula: C₃₃H₂₈N₂O₅
MDL Number: MFCD02682353
CAS Number: 1219348-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR