Fmoc-4-methyl-DL-tryptophan

Supplier: Matrix Scientific

042138-100MG 042138-1G
101854-622EA 289.23 USD
101854-622 101854-624
Fmoc-4-methyl-DL-tryptophan
Fmoc-4-methyl-DL-tryptophan

Hazard: Irritant

Formula: C₂₇H₂₄N₂O₄
MDL Number: MFCD09750499
CAS Number: 1219279-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR