Fmoc-1-methyl-DL-tryptophan
Supplier: Matrix Scientific
042137-100MG
042137-1G
101854-618EA
249.23
USD
101854-618
101854-620
Fmoc-1-methyl-DL-tryptophan
Fmoc-1-methyl-DL-tryptophan
Hazard: Irritant
Formula:
C₂₇H₂₄N₂O₄ |
MDL Number:
MFCD02682373 CAS Number: 138775-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...