Boc-DL-7-azatryptophan

Supplier: Matrix Scientific

Synonyms: 2-((tert-Butoxycarbonyl)amino)-3-(1H-pyrrolo[2,3-b]pyridin-3-yl)propanoic acid

042135-1G 042135-5G
101854-610EA 958.46 USD
101854-610 101854-612
Boc-DL-7-azatryptophan
Boc-DL-7-azatryptophan

Hazard: Irritant

Formula: C₁₅H₁₉N₃O₄
MW: 305.34 g/mol
MDL Number: MFCD02682350
CAS Number: 129423-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR