Boc-7-bromo-DL-tryptophan
Supplier: Matrix Scientific
042130-100MG
042130-1G
101854-590EA
326.15
USD
101854-590
101854-592
Boc-7-bromo-DL-tryptophan
Boc-7-bromo-DL-tryptophan
Hazard: Irritant
Formula:
C₁₆H₁₉BrN₂O₄ MW: 383.25 g/mol |
MDL Number:
MFCD09750490 CAS Number: 1219432-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...