Boc-6-methyl-DL-tryptophan
Supplier: Matrix Scientific
042128-100MG
042128-1G
101854-582EA
498.46
USD
101854-582
101854-584
Boc-6-methyl-DL-tryptophan
Boc-6-methyl-DL-tryptophan
Hazard: Irritant
Formula:
C₁₇H₂₂N₂O₄ |
MDL Number:
MFCD02682376 CAS Number: 446847-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...