Boc-6-fluoro-DL-tryptophan

Supplier: Matrix Scientific

042127-100MG 042127-1G
101854-578EA 275.38 USD
101854-578 101854-580
Boc-6-fluoro-DL-tryptophan
Boc-6-fluoro-DL-tryptophan

Hazard: Irritant

Formula: C₁₆H₁₉FN₂O₄
MW: 322.34 g/mol
MDL Number: MFCD02682360
CAS Number: 67308-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR