Boc-6-chloro-D-tryptophan

Supplier: Matrix Scientific

042126-100MG 042126-500MG
101854-574EA 772.31 USD
101854-574 101854-576
Boc-6-chloro-D-tryptophan
Boc-6-chloro-D-tryptophan

Hazard: Irritant

Formula: C₁₆H₁₉ClN₂O₄
MDL Number: MFCD09750493
CAS Number: 1217738-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR