Boc-6-chloro-DL-tryptophan

Supplier: Matrix Scientific

042125-100MG 042125-1G
101854-570EA 363.08 USD
101854-570 101854-572
Boc-6-chloro-DL-tryptophan
Boc-6-chloro-DL-tryptophan

Hazard: Irritant

Formula: C₁₆H₁₉ClN₂O₄
MDL Number: MFCD09750495
CAS Number: 1219193-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR