Boc-6-bromo-DL-tryptophan

Supplier: Matrix Scientific

042124-100MG 042124-1G
101854-566EA 326.15 USD
101854-566 101854-568
Boc-6-bromo-DL-tryptophan
Boc-6-bromo-DL-tryptophan

Hazard: Irritant

Formula: C₁₆H₁₉BrN₂O₄
MDL Number: MFCD09750488
CAS Number: 1104606-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR