Boc-5-methyl-DL-tryptophan
Supplier: Matrix Scientific
042123-100MG
042123-1G
101854-562EA
300
USD
101854-562
101854-564
Boc-5-methyl-DL-tryptophan
Boc-5-methyl-DL-tryptophan
Hazard: Irritant
Formula:
C₁₇H₂₂N₂O₄ MW: 318.38 g/mol |
MDL Number:
MFCD02682374 CAS Number: 142847-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...