Boc-5-methoxy-L-tryptophan

Supplier: Matrix Scientific

042122-250MG 042122-1G
101854-558EA 686.15 USD
101854-558 101854-560
Boc-5-methoxy-L-tryptophan
Boc-5-methoxy-L-tryptophan

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₅
MDL Number: MFCD09750497
CAS Number: 114903-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR