Boc-5-methoxy-2-methyl-DL-tryptophan
Supplier: Matrix Scientific
042120-100MG
042120-1G
101854-550EA
190.77
USD
101854-550
101854-552
Boc-5-methoxy-2-methyl-DL-tryptophan
Boc-5-methoxy-2-methyl-DL-tryptophan
Hazard: Irritant
Formula:
C₁₈H₂₄N₂O₅ |
MDL Number:
MFCD02682364 CAS Number: 1219410-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...