Boc-5-hydroxy-L-tryptophan
Supplier: Matrix Scientific
042119-1G
042119-5G
101854-546EA
412.31
USD
101854-546
101854-548
Boc-5-hydroxy-L-tryptophan
Boc-5-hydroxy-L-tryptophan
Hazard: Irritant
Formula:
C₁₆H₂₀N₂O₅ |
MDL Number:
MFCD00270732 CAS Number: 119768-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...