Boc-5-hydroxy-DL-tryptophan

Supplier: Matrix Scientific

042118-250MG 042118-1G
101854-542EA 1053.85 USD
101854-542 101854-544
Boc-5-hydroxy-DL-tryptophan
Boc-5-hydroxy-DL-tryptophan

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₅
MW: 320.35 g/mol
MDL Number: MFCD02682362
CAS Number: 185525-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR