Boc-5-chloro-DL-tryptophan

Supplier: Matrix Scientific

042116-100MG 042116-1G
101854-534EA 363.08 USD
101854-534 101854-536
Boc-5-chloro-DL-tryptophan
Boc-5-chloro-DL-tryptophan

Hazard: Irritant

Formula: C₁₆H₁₉ClN₂O₄
MW: 338.79 g/mol
MDL Number: MFCD09264328
CAS Number: 361576-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR