Boc-5-bromo-DL-tryptophan

Supplier: Matrix Scientific

042115-100MG 042115-1G
101854-530EA 326.15 USD
101854-530 101854-532
Boc-5-bromo-DL-tryptophan
Boc-5-bromo-DL-tryptophan

Hazard: Irritant

Formula: C₁₆H₁₉BrN₂O₄
MDL Number: MFCD02682356
CAS Number: 67308-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR