Boc-5-benzyloxy-DL-tryptophan

Supplier: Matrix Scientific

042114-250MG 042114-1G
101854-526EA 363.08 USD
101854-526 101854-528
Boc-5-benzyloxy-DL-tryptophan
Boc-5-benzyloxy-DL-tryptophan

Hazard: Irritant

Formula: C₂₃H₂₆N₂O₅
MDL Number: MFCD02682352
CAS Number: 1219197-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR