Boc-1-methyl-DL-tryptophan

Supplier: Matrix Scientific

042112-100MG 042112-1G
101854-518EA 380 USD
101854-518 101854-520
Boc-1-methyl-DL-tryptophan
Boc-1-methyl-DL-tryptophan

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₄
MDL Number: MFCD02682372
CAS Number: 185527-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR