Fmoc-DL-2-amino-4-trityloxybutanoic acid

Supplier: Matrix Scientific

042110-1G 042110-5G
101854-510EA 326.15 USD
101854-510 101854-512
Fmoc-DL-2-amino-4-trityloxybutanoic acid
Fmoc-DL-2-amino-4-trityloxybutanoic acid

Hazard: Irritant

Formula: C₃₈H₃₃NO₅
MW: 583.69 g/mol
MDL Number: MFCD04974563
CAS Number: 1396967-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR