Fmoc-L-HomoSer(Trt)-OH
Supplier: Matrix Scientific
Synonyms:
Fmoc-HomoSer(Trt)-OH, Fmoc-HoSer(Trt)-OH, Fmoc-Hse(Trt)-OH
042107-1G
042107-5G
101854-498EA
253.85
USD
101854-498
101854-500
Fmoc-L-HomoSer(Trt)-OH
Fmoc-L-HomoSer(Trt)-OH
Hazard: Irritant
Formula:
C₃₈H₃₃NO₅ |
MDL Number:
MFCD00270544 CAS Number: 111061-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...