Fmoc-2-amino-3-methoxypropionic acid

Supplier: Matrix Scientific

042106-100MG 042106-1G 042106-5G
101854-492EA 224.62 USD
101854-492 101854-494 101854-496
Fmoc-2-amino-3-methoxypropionic acid
Fmoc-2-amino-3-methoxypropionic acid

Hazard: Irritant

Formula: C₁₉H₁₉NO₅
MDL Number: MFCD02682606
CAS Number: 215189-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR