Fmoc-2-amino-3-methoxybutanoic acid

Supplier: Matrix Scientific

042105-100MG 042105-250MG
101854-488EA 823.08 USD
101854-488 101854-490
Fmoc-2-amino-3-methoxybutanoic acid
Fmoc-2-amino-3-methoxybutanoic acid

Hazard: Irritant

Formula: C₂₀H₂₁NO₅
MDL Number: MFCD02682602
CAS Number: 1396995-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR