Fmoc-2-amino-3-hydroxypentanoic acid
Supplier: Matrix Scientific
042104-100MG
042104-250MG
101854-484EA
547.69
USD
101854-484
101854-486
Fmoc-2-amino-3-hydroxypentanoic acid
Fmoc-2-amino-3-hydroxypentanoic acid
Hazard: Irritant
Formula:
C₂₀H₂₁NO₅ |
MDL Number:
MFCD02682590 CAS Number: 1219207-99-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...