Fmoc-(S)-2-amino-3-methoxypropionic acid

Supplier: Matrix Scientific

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methoxypropanoic acid, Fmoc-Ser(Me)-OH

042103-1G 042103-5G
101854-480EA 150.77 USD
101854-480 101854-482
Fmoc-(S)-2-amino-3-methoxypropionic acid
Fmoc-(S)-2-amino-3-methoxypropionic acid

Hazard: Irritant

Formula: C₁₉H₁₉NO₅
MW: 341.37 g/mol
MDL Number: MFCD02682604
CAS Number: 159610-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR