Fmoc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid

Supplier: Matrix Scientific

Synonyms: Fmoc-Val(3-OH)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxy-3-methylbutanoic acid

042102-100MG 042102-500MG
101854-476EA 120 USD
101854-476 101854-478
Fmoc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid
Fmoc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid

Hazard: Irritant

Formula: C₂₀H₂₁NO₅
MDL Number: MFCD04974410
CAS Number: 1217603-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR