Fmoc-(2S,3S)-3-amino-2-hydroxy-5-methylhexanoic acid

Supplier: Matrix Scientific

042095-100MG 042095-500MG 042095-1G
101854-444EA 661.54 USD
101854-444 101854-446 101854-448
Fmoc-(2S,3S)-3-amino-2-hydroxy-5-methylhexanoic acid
Fmoc-(2S,3S)-3-amino-2-hydroxy-5-methylhexanoic acid

Hazard: Irritant

Formula: C₂₂H₂₅NO₅
MDL Number: MFCD04974449
CAS Number: 361161-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR