Fmoc-(2S,3S)-2-amino-3-methoxybutanoic acid
Supplier: Matrix Scientific
042094-100MG
042094-250MG
101854-440EA
823.08
USD
101854-440
101854-442
Fmoc-(2S,3S)-2-amino-3-methoxybutanoic acid
Fmoc-(2S,3S)-2-amino-3-methoxybutanoic acid
Hazard: Irritant
Formula:
C₂₀H₂₁NO₅ |
MDL Number:
MFCD02682600 CAS Number: 1279029-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...