Fmoc-(2S,3RS)-2-amino-3-hydroxy-4-methylpentanoicacid
Supplier: Matrix Scientific
042092-100MG
042092-250MG
101854-432EA
686.15
USD
101854-432
101854-434
Fmoc-(2S,3RS)-2-amino-3-hydroxy-4-methylpentanoicacid
Fmoc-(2S,3RS)-2-amino-3-hydroxy-4-methylpentanoicacid
Hazard: Irritant
Formula:
C₂₁H₂₃NO₅ |
MDL Number:
MFCD02682587 CAS Number: 1217516-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...