Fmoc-(2R,3S)-3-amino-2-hydroxy-5-methylhexanoic acid

Supplier: Matrix Scientific

042090-100MG 042090-500MG 042090-1G
101854-420EA 661.54 USD
101854-420 101854-422 101854-424
Fmoc-(2R,3S)-3-amino-2-hydroxy-5-methylhexanoic acid
Fmoc-(2R,3S)-3-amino-2-hydroxy-5-methylhexanoic acid

Hazard: Irritant

Formula: C₂₂H₂₅NO₅
MDL Number: MFCD04974451
CAS Number: 361161-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR