Fmoc-(2R,3S)-2-amino-3-hydroxy-4-methylpentanoicacid

Supplier: Matrix Scientific

042089-100MG 042089-250MG
101854-416EA 823.08 USD
101854-416 101854-418
Fmoc-(2R,3S)-2-amino-3-hydroxy-4-methylpentanoicacid
Fmoc-(2R,3S)-2-amino-3-hydroxy-4-methylpentanoicacid

Hazard: Irritant

Formula: C₂₁H₂₃NO₅
MDL Number: MFCD02682583
CAS Number: 1217833-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR