Fmoc-(2R,3R)-3-amino-2-hydroxy-5-methylhexanoic acid

Supplier: Matrix Scientific

042088-100MG 042088-500MG 042088-1G
101854-410EA 661.54 USD
101854-410 101854-412 101854-414
Fmoc-(2R,3R)-3-amino-2-hydroxy-5-methylhexanoic acid
Fmoc-(2R,3R)-3-amino-2-hydroxy-5-methylhexanoic acid

Hazard: Irritant

Formula: C₂₂H₂₅NO₅
MDL Number: MFCD02682581
CAS Number: 1217603-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR