Boc-L-α-methylserine

Supplier: Matrix Scientific

042087-100MG 042087-1G
101854-406EA 115.38 USD
101854-406 101854-408
Boc-L-α-methylserine
Boc-L-alpha-methylserine

Hazard: Irritant

Formula: C₉H₁₇NO₅
MW: 219.24 g/mol
MDL Number: MFCD02682593
CAS Number: 84311-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR