Boc-DL-Hse(Bzl)-OH

Supplier: Matrix Scientific

042086-1G 042086-5G
101854-402EA 326.15 USD
101854-402 101854-404
Boc-DL-Hse(Bzl)-OH
Boc-DL-Hse(Bzl)-OH

Hazard: Irritant

Formula: C₁₆H₂₃NO₅
MW: 309.37 g/mol
MDL Number: MFCD04974562
CAS Number: 876617-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR