Boc-4-amino-3-hydroxybutanoic acid

Supplier: Matrix Scientific

042084-1G 042084-5G
101854-394EA 436.92 USD
101854-394 101854-396
Boc-4-amino-3-hydroxybutanoic acid
Boc-4-amino-3-hydroxybutanoic acid

Hazard: Irritant

Formula: C₉H₁₇NO₅
MDL Number: MFCD02682574
CAS Number: 69489-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR