Boc-L-HomoSer(Bzl)-OH

Supplier: Matrix Scientific

Synonyms: Boc-Hse(Bzl)-OH, Boc-HomoSer(Bzl)-OH

042083-1G 042083-5G
101854-390EA 326.15 USD
101854-390 101854-392
Boc-L-HomoSer(Bzl)-OH
Boc-L-HomoSer(Bzl)-OH

Hazard: Irritant

Formula: C₁₆H₂₃NO₅
MW: 309.4 g/mol
MDL Number: MFCD00270497
CAS Number: 59408-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR