Boc-O-methyl-L-serine

Supplier: Matrix Scientific

Synonyms: Boc-2-amino-3-methoxypropionic acid, Boc-Ser(Me)-OH, N-tert-Butoxycarbonyl-O-methyl-L-serine, (2S)-2-Amino-3-methoxypropanoic acid, N-BOC protected

042082-100MG 042082-1G 042082-5G
101854-384EA 224.62 USD
101854-384 101854-386 101854-388
Boc-O-methyl-L-serine
Boc-O-methyl-L-serine

Hazard: Irritant

Formula: C₉H₁₇NO₅
MW: 219.237 g/mol
MDL Number: MFCD00153314
CAS Number: 51293-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR