Boc-2-amino-3-methoxybutanoic acid

Supplier: Matrix Scientific

042081-100MG 042081-250MG
101854-380EA 823.08 USD
101854-380 101854-382
Boc-2-amino-3-methoxybutanoic acid
Boc-2-amino-3-methoxybutanoic acid

Hazard: Irritant

Formula: C₁₀H₁₉NO₅
MDL Number: MFCD02682601
CAS Number: 1396972-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR