Boc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid
Supplier: Matrix Scientific
Synonyms:
Boc-Val(3-OH)-OH, 3-Hydroxy-L-valine, N-BOC protected, (S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxy-3-methylbutanoic acid
042078-500MG
042078-1G
101854-370EA
469.23
USD
101854-370
102551-302
Boc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid
Boc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid
Hazard: Irritant
Formula:
C₁₀H₁₉NO₅ MW: 233.27 g/mol |
MDL Number:
MFCD03094792 CAS Number: 102507-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...