(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid

Supplier: Matrix Scientific

Synonyms: Boc-Ser(Bzl)-OH, N-Boc-O-benzyl-L-serine

042076-1G 042076-5G
101854-360EA 56.92 USD
101854-360 101854-362
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid

Hazard: Irritant

Formula: C₁₅H₂₁NO₅
MDL Number: MFCD00066063
CAS Number: 23680-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR