Boc-(RS)-2-amino-3-hydroxy-3-methylbutanoic acid

Supplier: Matrix Scientific

042075-100MG 042075-1G
101854-356EA 138.46 USD
101854-356 101854-358
Boc-(RS)-2-amino-3-hydroxy-3-methylbutanoic acid
Boc-(RS)-2-amino-3-hydroxy-3-methylbutanoic acid

Hazard: Irritant

Formula: C₁₀H₁₉NO₅
MDL Number: MFCD02682578
CAS Number: 105504-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR