O-Benzyl-N-(tert-butoxycarbonyl)-D-serine
Supplier: Matrix Scientific
Synonyms:
Boc-D-Ser(Bzl)-OH, N-Boc-O-benzyl-D-serine
042073-1G
042073-5G
101854-348EA
36.92
USD
101854-348
101854-350
O-Benzyl-N-(tert-butoxycarbonyl)-D-serine
O-Benzyl-N-(tert-butoxycarbonyl)-D-serine
Hazard: Irritant
Formula:
C₁₅H₂₁NO₅ MW: 295.34 g/mol |
MDL Number:
MFCD00038248 CAS Number: 47173-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...