Boc-statine

Supplier: Matrix Scientific

042072-100MG 042072-1G
101854-344EA 275.38 USD
101854-344 101854-346
Boc-statine
Boc-statine

Hazard: Irritant

Formula: C₁₃H₂₅NO₅
MW: 275.345 g/mol
MDL Number: MFCD00063180
CAS Number: 58521-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR